Learn More
Bellco Glass, Inc. Polypropylene Griffin Grad Beaker,50mL
SDP
Catalog No. 501945715
Encompass
These beakers are useful for general laboratory procedures and are autoclaveable. They have blue imprinted graduations to aid in viewing and dispensing liquids. Material: polypropylene Size: 50ml x 10mL Dimensions: 40x60mm
Catalog No. 50-194-5715 Supplier Bellco Glass, Inc. Supplier No. 562300050
These beakers are useful for general laboratory procedures and are autoclaveable. They have blue imprinted graduations to aid in viewing and dispensing liquids. Material: polypropylene Size: 50ml x 10mL Dimensions: 40x60mm