Learn More
Bellco Glass, Inc. Polypropylene Griffin Grad Beaker,600mL
SDP
Catalog No. 501945721
Encompass
These beakers are useful for general laboratory procedures and are autoclaveable. They have blue imprinted graduations to aid in viewing and dispensing liquids. Material: polypropylene Size: 600ml x 100mL Dimensions: 91x125mm
Catalog No. 50-194-5721 Supplier Bellco Glass, Inc. Supplier No. 562300600
These beakers are useful for general laboratory procedures and are autoclaveable. They have blue imprinted graduations to aid in viewing and dispensing liquids. Material: polypropylene Size: 600ml x 100mL Dimensions: 91x125mm