Learn More
Molecular Research Center PP MICROFUGE TUBES 2ML STL
SDP
Catalog No. NC9521468
Encompass
DISPOSABLE POLYPROPYLENE MICROFUGE TUBES,2 ml sterile 200/box,Made of ultra-clear polypropylene,are tested in RNA,DNA and protein isolation using TRI Reagents,and withstand centrifugal forces of 12,000 g
Catalog No. NC9521468 Supplier Molecular Research Center Supplier No. PP 132
DISPOSABLE POLYPROPYLENE MICROFUGE TUBES,2 ml sterile 200/box,Made of ultra-clear polypropylene,are tested in RNA,DNA and protein isolation using TRI Reagents,and withstand centrifugal forces of 12,000 g