Learn More
Accuflow Systems Mini TC x 3/8" ID Polypropylene Bio Barb Adapters 10/pack
Supplier:  Accuflow Systems PPMINITC0375BB
Encompass_Preferred
Mini TC x 3/8" ID Polypropylene Bio Barb Adapters with true-bore ID are designed for use in GMP Manufacturing and laboratories. These Bio Barb adapters can be autoclavable as well as gama irradiated. Constructed of USP Class VI Polypropylene. Designed for Sanitary Tri clamp applications. Max working pressure, 150psi @ 70? F, Pack of 10
Catalog No. 50-110-8266