Learn More
Bellco Glass, Inc. PP Pestle Only,0.5mL,Assorted Colors
SDP
Catalog No. 501945178
Encompass_Preferred
Pp pestle w/ assorted colors 0.5ml. Homogenize cells or resuspend precipitates such as protein or DNA pellets. Certified free of RNase, DNase, DNA, PCR inhibitors and tested pyrogen-free.
Catalog No. 50-194-5178 Supplier Bellco Glass, Inc. Supplier No. 198000005
Pp pestle w/ assorted colors 0.5ml. Homogenize cells or resuspend precipitates such as protein or DNA pellets. Certified free of RNase, DNase, DNA, PCR inhibitors and tested pyrogen-free.