Learn More
Bellco Glass, Inc. PP Pestle Only,1.5mL, Assorted Colors
SDP
Catalog No. 501945179
Encompass_Preferred
Pp pestle only w/ assorted colors 1.5ml. Homogenize cells or resuspend precipitates such as protein or DNA pellets. Certified free of RNase, DNase, DNA, PCR inhibitors and tested pyrogen-free.
Catalog No. 50-194-5179 Supplier Bellco Glass, Inc. Supplier No. 198000015
Pp pestle only w/ assorted colors 1.5ml. Homogenize cells or resuspend precipitates such as protein or DNA pellets. Certified free of RNase, DNase, DNA, PCR inhibitors and tested pyrogen-free.