Learn More
Cole-Parmer Scienceware H14660-0065 polypropylene powder funnel, 70 mL
Catalog No. NC0663075 Shop All Cole-Parmer Products
Encompass_Preferred
Polypropylene funnels have 60°> wall incline. Use for transferring powders. Parallel stem minimizes bridging of powder. Bel-Art.
Catalog No. NC0663075 Supplier Cole-Parmer Supplier No. 0612310
Polypropylene funnels have 60°> wall incline. Use for transferring powders. Parallel stem minimizes bridging of powder. Bel-Art.