Learn More
Premier Lab Supply Polypropylene 4.0μm (0.16mil) Sheets – PN# TF-240-345
SDP
Catalog No. NC2162800
Encompass_Preferred
Polypropylene film 4.0μm (0.16mil) sheets with Dimensions: 3” x 3” (76 x 76mm) (PN# TF-240-345). General purpose film with excellent X-ray transmission and chemical resistance, and lot number tracking system. 500 per box.
This item is not returnable. View return policy
Catalog No. NC2162800 Supplier Premier Lab Supply Supplier No. TF240345
Polypropylene film 4.0μm (0.16mil) sheets with Dimensions: 3” x 3” (76 x 76mm) (PN# TF-240-345). General purpose film with excellent X-ray transmission and chemical resistance, and lot number tracking system. 500 per box.