Learn More
Bellco Glass, Inc. Individually calibrated to deliver at 20C. Durable ring marks resilient wear. Certified Class B with individual ring marking per DIN 122664. Supplied with screw cap. Volume: 1l Accuracy +/- ml: 0.60 Height: 310mm Thread: 32
SDP
Catalog No. 501945706
Encompass
Individually calibrated to deliver at 20C. Durable ring marks resilient wear. Certified Class B with individual ring marking per DIN 122664. Supplied with screw cap. Volume: 1l Accuracy +/- ml: 0.60 Height: 310mm Thread: 32
Catalog No. 50-194-5706 Supplier Bellco Glass, Inc. Supplier No. 562201000
Individually calibrated to deliver at 20C. Durable ring marks resilient wear. Certified Class B with individual ring marking per DIN 122664. Supplied with screw cap. Volume: 1l Accuracy +/- ml: 0.60 Height: 310mm Thread: 32