Learn More
Bellco Glass, Inc. Individually calibrated to deliver at 20C. Durable ring marks resilient wear. Certified Class B with individual ring marking per DIN 122664. Supplied with screw cap. Volume: 100ml Accuracy +/- ml: 0.16 Height: 180mm Thread: 18
SDP
Catalog No. 501945703
Encompass
Individually calibrated to deliver at 20C. Durable ring marks resilient wear. Certified Class B with individual ring marking per DIN 122664. Supplied with screw cap. Volume: 100ml Accuracy +/- ml: 0.16 Height: 180mm Thread: 18
Catalog No. 50-194-5703 Supplier Bellco Glass, Inc. Supplier No. 562200100
Individually calibrated to deliver at 20C. Durable ring marks resilient wear. Certified Class B with individual ring marking per DIN 122664. Supplied with screw cap. Volume: 100ml Accuracy +/- ml: 0.16 Height: 180mm Thread: 18