Learn More
Bellco Glass, Inc. Individually calibrated to deliver at 20C. Durable ring marks resilient wear. Certified Class B with individual ring marking per DIN 122664. Supplied with screw cap. Volume ml: 25 ml Accuracy +/- ml : 0.06 Height: 115mm Thread gl: 18
SDP
Catalog No. 501945701
Encompass
Individually calibrated to deliver at 20C. Durable ring marks resilient wear. Certified Class B with individual ring marking per DIN 122664. Supplied with screw cap. Volume ml: 25 ml Accuracy +/- ml : 0.06 Height: 115mm Thread gl: 18
Catalog No. 50-194-5701 Supplier Bellco Glass, Inc. Supplier No. 562200025
Individually calibrated to deliver at 20C. Durable ring marks resilient wear. Certified Class B with individual ring marking per DIN 122664. Supplied with screw cap. Volume ml: 25 ml Accuracy +/- ml : 0.06 Height: 115mm Thread gl: 18