Learn More
Bellco Glass, Inc. Individually calibrated to deliver at 20C. Durable ring marks resilient wear. Certified Class B with individual ring marking per DIN 122664. Supplied with screw cap. Volume: 500ml Accuracy +/- ml: 0.40 Height: 270mm Thread: 25
SDP
Catalog No. 501945705
Encompass
Individually calibrated to deliver at 20C. Durable ring marks resilient wear. Certified Class B with individual ring marking per DIN 122664. Supplied with screw cap. Volume: 500ml Accuracy +/- ml: 0.40 Height: 270mm Thread: 25
Catalog No. 50-194-5705 Supplier Bellco Glass, Inc. Supplier No. 562200500
Individually calibrated to deliver at 20C. Durable ring marks resilient wear. Certified Class B with individual ring marking per DIN 122664. Supplied with screw cap. Volume: 500ml Accuracy +/- ml: 0.40 Height: 270mm Thread: 25