Learn More
Bellco Glass, Inc. Individually calibrated to deliver at 20C. Durable ring marks resilient wear. Certified Class B with individual ring marking per DIN 122664. Supplied with screw cap. Volume ml: 50ml Accuracy +/- ml: 0.10 Height: 150mm Thread gl: 18
SDP
Catalog No. 501945702
Encompass_Preferred
Individually calibrated to deliver at 20C. Durable ring marks resilient wear. Certified Class B with individual ring marking per DIN 122664. Supplied with screw cap. Volume ml: 50ml Accuracy +/- ml: 0.10 Height: 150mm Thread gl: 18
Catalog No. 50-194-5702 Supplier Bellco Glass, Inc. Supplier No. 562200050
Individually calibrated to deliver at 20C. Durable ring marks resilient wear. Certified Class B with individual ring marking per DIN 122664. Supplied with screw cap. Volume ml: 50ml Accuracy +/- ml: 0.10 Height: 150mm Thread gl: 18