Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Polypropylene, 45 Elbow, Sanitary Clamp Union, 1 1/2"" Clamp x 1 1/2"" Clamp
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3180653/DEL
Encompass
Tri-Clamp 45-degree elbow adapters allow for secure connection of sanitary lines when a 45-degree angle is required. Though both materials may be autoclaved, PVDF tolerates high temperatures better than PP does and will perform more consistently after repeated autoclaving. Clamps and gaskets required for a complete system are sold separately.
Catalog No. NC1575392
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.