Learn More
AVANTOR MASTERFLEX (VWR LLC) Sanitary Tee, Polypropylene, 1"" Ladish Clamp Size
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3180662/DEL
Encompass
Sanitary Tee, Polypropylene, 1" Clamp, Maintain sanitary conditions while combining or dividing flowMeet USP Class VI for pharmaceutical use and FDA, USDA, and 3A standards for food contactAll fittings are compatible with Tri-Clamp fittingsExcellent chemical resistance; low leachable characteristics
Catalog No. NC1494721