Learn More
AVANTOR MASTERFLEX (VWR LLC) Sanitary Tee, 3/4"" Mini Clamp Size, Polypropylene
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3180661/DEL
Encompass
Sanitary Tee, Polypropylene, 3/4" Mini Clamp, Maintain sanitary conditions while combining or dividing flowMeet USP Class VI for pharmaceutical use and FDA, USDA, and 3A standards for food contactAll fittings are compatible with Tri-Clamp fittingsExcellent chemical resistance; low leachable characteristics,
Catalog No. NC0907987