Learn More
Bellco Glass, Inc. PP Snap Cap W/10mil PTFE,11mm
SDP
Catalog No. 501945400
Encompass_Preferred
Used for snap thread vials. Caps may be snapped on and off by hand which eliminated the need for a metal crimping and decapping tool. MADE IN THE USA. Size: 11mm Septa material: ptfe
Catalog No. 50-194-5400 Supplier Bellco Glass, Inc. Supplier No. 207003100
Used for snap thread vials. Caps may be snapped on and off by hand which eliminated the need for a metal crimping and decapping tool. MADE IN THE USA. Size: 11mm Septa material: ptfe