Learn More
ANALYTICS SHOP USA LP PP Storage Box for 4ml vials or 4ml shell, vials, red, cover, (130x130x52mm), 49 cavities
SDP
Catalog No. NC3316236
Encompass_Preferred
PP Storage Boxes-Safe standing position on the laboratory gable and during transport due to specific cavities related to the vial diameter.-Ideal for space-saving storage in fridges, as the transparent lid prevents condensations on the closures and thus avoids a possible contamination in the cooling unit. -Temperature resistant from -80 degrees up to +100 degrees.-Alpha Numeric Coding (1.5ml, 4ml) for clear sample identification.-Unbreakable polypropylene bottom and lid, stackable.-Chemically resistant and fairly robust; autoclavable.-With alphanumeric coding at the cavities
Catalog No. NC3316236 Supplier ANALYTICS SHOP USA LP Supplier No. AAV26323537
PP Storage Boxes-Safe standing position on the laboratory gable and during transport due to specific cavities related to the vial diameter.-Ideal for space-saving storage in fridges, as the transparent lid prevents condensations on the closures and thus avoids a possible contamination in the cooling unit. -Temperature resistant from -80 degrees up to +100 degrees.-Alpha Numeric Coding (1.5ml, 4ml) for clear sample identification.-Unbreakable polypropylene bottom and lid, stackable.-Chemically resistant and fairly robust; autoclavable.-With alphanumeric coding at the cavities