Learn More
Chemglass Life Sciences Syringe, Polypropylene, Luer Lock, 20mL, Non-Sterile, Bulk
SDP
Supplier:  Chemglass Life Sciences CGE308304
Encompass
All polypropylene syringes with a Luer Lock. The plastic plunger has a smooth draw and positive safety stop to prevent accidental spills. Smooth draw.
Catalog No. 50-189-7593