Learn More
Fisherbrand™ PP TPE Gasket Closure, 83B, 2-Port, 1/4 in.
Catalog No. 14388173 Shop All Fisherbrand Products
Change view
Click to view available options
Thread Size (Metric):
43 mm
Catalog No. Thread Size (Metric)
14-388-173 43 mm
1 options
Catalog No. 14-388-173
Add to Cart
Add to Cart
Nalgene Heavy-Duty Vacuum Carboys and Bottles
  • Maintain the integrity of a container and its contents with FBPP, TPE Gskt, Clsr, 83B, 2port, 1/4 in.
  • A convenient fluid transfer system, this ported 83B closure features a quick-disconnect fitting with tubulation for attachment to tubing of your choice.
  • Fittings have tubulation both in and outside of the closure, and socket fittings in the cap have internal valves that seal when a tubulation fitting is removed to maintain container seal integrity.
  • Ready-made alternative to cumbersome in house filling/dispensing solutions
  • Assures container integrity with shut off valve style quick-disconnect fittings
  • 83B closure features inside/outside barbed fittings
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Material Polypropylene
Sterility Non-sterile
Thread Size (Metric) 43 mm
Neck Finish 43 mm
Autoclavable Not Autoclavable
Disposable Reusable
Product Type Screw Closure and Gasket