Learn More
Beckman Coulter 15 mL, Open-Top Thickwall Open-Top Tube, 18 x 100mm, 100/pk
Catalog No. NC9253707
Encompass_Preferred
Polypropylene Tubes For JA-20.1 Rotor, Quantity 100, Volume per Tube 15mL, Size 18 x 98mm
Catalog No. NC9253707 Supplier Beckman Coulter Supplier No. 342082
Polypropylene Tubes For JA-20.1 Rotor, Quantity 100, Volume per Tube 15mL, Size 18 x 98mm