Learn More
Cole-Parmer Cole-Parmer High-density polyethylene utility tray, 17 3/4" x 14" x 4"
Catalog No. NC9588221 Shop All Cole-Parmer Products
Encompass_Preferred
These durable, 3/16'' thick high-density polyethylene trays are NSF-certified and meet USDA requirements.
Catalog No. NC9588221 Supplier Cole-Parmer Supplier No. 0671010
These durable, 3/16'' thick high-density polyethylene trays are NSF-certified and meet USDA requirements.