Learn More
Microsolv Technology Corporation Pp Vials 2Ml Clear
Catalog No. 5001452190
Encompass_Preferred
Vials, Screw Top, Plastic. Translucent, 2ml, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52190 Supplier Microsolv Technology Corporation Supplier No. 9502SPPCLEAR
Vials, Screw Top, Plastic. Translucent, 2ml, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. MicroSolv Brand. 100/PK.