Learn More
Microsolv Technology Corporation Pp Vials And Caps Preslit
Catalog No. 5001452188
Encompass_Preferred
Vial & Cap Kit Includes 100 EA 2ml, Screw Top, Clear Polypropylene Autosampler Vials with Fill lines and 100 EA matching Screw Caps with Silicone Rubber / PTFE Pre-Slit Septa, bonded in the Blue Caps, MicroSolv Brand Easy Purchase Pack. 1/PK.
Catalog No. 50-014-52188 Supplier Microsolv Technology Corporation Supplier No. 9502SPP3CP
Vial & Cap Kit Includes 100 EA 2ml, Screw Top, Clear Polypropylene Autosampler Vials with Fill lines and 100 EA matching Screw Caps with Silicone Rubber / PTFE Pre-Slit Septa, bonded in the Blue Caps, MicroSolv Brand Easy Purchase Pack. 1/PK.