Learn More
Bellco Glass, Inc. Polypropylene Volumetric Flask W/Polypropylene Stopper,1000mL Class B
SDP
Catalog No. 501945712
Encompass
Individually calibrated to deliver at 20C. Durable ring marks resilient wear. Supplied with standard ground polypropylene stopper. Certified Class B tolerance per DIN ISO 1042. Translucent with individual ring mark. Volume: 1l Accuracy +/- ml: 0.60 Height: 310mm Stopper size: 24/29
Catalog No. 50-194-5712 Supplier Bellco Glass, Inc. Supplier No. 562211000
Individually calibrated to deliver at 20C. Durable ring marks resilient wear. Supplied with standard ground polypropylene stopper. Certified Class B tolerance per DIN ISO 1042. Translucent with individual ring mark. Volume: 1l Accuracy +/- ml: 0.60 Height: 310mm Stopper size: 24/29