Learn More
Bellco Glass, Inc. Polypropylene Volumetric Flask W/Polypropylene Stopper,100mL Class B
SDP
Catalog No. 501945709
Encompass
Individually calibrated to deliver at 20C. Durable ring marks resilient wear. Supplied with standard ground polypropylene stopper. Certified Class B tolerance per DIN ISO 1042. Translucent with individual ring mark. Volume: 100ml Accuracy +/- ml: 0.16 Height: 180mm Stopper size: 14/23
Catalog No. 50-194-5709 Supplier Bellco Glass, Inc. Supplier No. 562210100
Individually calibrated to deliver at 20C. Durable ring marks resilient wear. Supplied with standard ground polypropylene stopper. Certified Class B tolerance per DIN ISO 1042. Translucent with individual ring mark. Volume: 100ml Accuracy +/- ml: 0.16 Height: 180mm Stopper size: 14/23