Learn More
Bellco Glass, Inc. Polypropylene Volumetric Flask W/Polypropylene Stopper,25mL Class B
SDP
Catalog No. 501945707
Encompass
Individually calibrated to deliver at 20C. Durable ring marks resilient wear. Supplied with standard ground polypropylene stopper. Certified Class B tolerance per DIN ISO 1042. Translucent with individual ring mark. Volume: 25ml Accuracy +/- ml: 0.06 Height: 115mm Stopper size: 10/19
Catalog No. 50-194-5707 Supplier Bellco Glass, Inc. Supplier No. 562210025
Individually calibrated to deliver at 20C. Durable ring marks resilient wear. Supplied with standard ground polypropylene stopper. Certified Class B tolerance per DIN ISO 1042. Translucent with individual ring mark. Volume: 25ml Accuracy +/- ml: 0.06 Height: 115mm Stopper size: 10/19