Learn More
Bellco Glass, Inc. Polypropylene Volumetric Flask W/Polypropylene Stopper,500mL Class B
SDP
Catalog No. 501945711
Encompass
Individually calibrated to deliver at 20C. Durable ring marks resilient wear. Supplied with standard ground polypropylene stopper. Certified Class B tolerance per DIN ISO 1042. Translucent with individual ring mark. Volume: 500ml Accuracy +/- ml: 0.40 Height: 270mm Stopper size: 19/26
Catalog No. 50-194-5711 Supplier Bellco Glass, Inc. Supplier No. 562210500
Individually calibrated to deliver at 20C. Durable ring marks resilient wear. Supplied with standard ground polypropylene stopper. Certified Class B tolerance per DIN ISO 1042. Translucent with individual ring mark. Volume: 500ml Accuracy +/- ml: 0.40 Height: 270mm Stopper size: 19/26