Learn More
Bellco Glass, Inc. Polypropylene Volumetric Flask W/Polypropylene Stopper,50mL Class B
SDP
Catalog No. 501945708
Encompass
Individually calibrated to deliver at 20C. Durable ring marks resilient wear. Supplied with standard ground polypropylene stopper. Certified Class B tolerance per DIN ISO 1042. Translucent with individual ring mark. Volume:50ml Accuracy +/- ml: 0.10 Height: 150mm Stopper size: 10/19
Catalog No. 50-194-5708 Supplier Bellco Glass, Inc. Supplier No. 562210050
Individually calibrated to deliver at 20C. Durable ring marks resilient wear. Supplied with standard ground polypropylene stopper. Certified Class B tolerance per DIN ISO 1042. Translucent with individual ring mark. Volume:50ml Accuracy +/- ml: 0.10 Height: 150mm Stopper size: 10/19