Learn More
Bellco Glass, Inc. PP Yellow Screw Cap,45mm Solid
SDP
Catalog No. 501945762
Encompass_Preferred
Molded from Polypropylene with inner ring that seals into the neck of the bottle. They are autoclavable to 140 degrees Celsius. MADE IN THE USA Size: 45mm Color: yellow
Catalog No. 50-194-5762 Supplier Bellco Glass, Inc. Supplier No. 563700048
Molded from Polypropylene with inner ring that seals into the neck of the bottle. They are autoclavable to 140 degrees Celsius. MADE IN THE USA Size: 45mm Color: yellow