Learn More
Selleck Chemical LLC PP1 10mg
Supplier: Selleck Chemical LLC S706010MG
PP1 (AGL 1872, EI 275) is a potent and selective Src inhibitor for Lck/Fyn with IC50 of 5 nM/ 6 nM. *For Research & Development use only. Product is not intended for drug, household, or human consumption.