Learn More
AdipoGen PP1
SDP
Supplier:  AdipoGen AGCR13562M001
Encompass_Preferred
Chemical. CAS 172889-26-8. Formula C16H19N5. MW 281.4. Highly potent and selective Scr family tyrosine kinase inhibitor. Shows anti-Ras cancer potential by blocking Ras-induced activation of PAK1. Antitumor compound. RIP2 inhibitor. Blocks TGF-beta-mediated cellular responses.
Catalog No. 50-256-2288