Learn More
Enzo Life Sciences PP1 (1mg). CAS: 172889-26-8
SDP
Supplier:  Enzo Life Sciences BMLEI2750001
Encompass_Preferred
A highly potent and selective inhibitor of Src family tyrosine kinases, IC50=5 nM for p56lck; p59fynT=6 nM; p60src=170 nM. Inhibition of Src family kinases by PP1 has been shown to: prevent a-thrombin-stimulated Akt phosphorylation, delay PAF-mediated STAT3 phosphorylation, block Zn2+-induced Ras activation, block adhesion-dependent tyrosine phosphorylation of p105HEF1 5, and inhibit HLA-DR-induced tyrosine phosphorylation without affecting the activation of ERK1/2. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml), slightly soluble in 100% ethanol.
. Long Term Storage: -20°C.
Catalog No. 50-201-0656