Learn More
Eurofins Discoverx PP1α, 250 µg. (14-595M)
Supplier:  Eurofins Discoverx 14595M
Encompass
The PP1α product is purified is untagged recombinant PP1α enzyme. This enzyme is full length. It retains its binding and enzymatic activity and is intended for use in phosphatase assays. This enzyme is provided as 250 μg of 0.63 mg/mL frozen solution. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9024