Learn More
Medchemexpress LLC PP1 (AGL 1872; EI 275) | 172889-26-8 | 99.6% | 281.36 | 50 MG
SDP
Catalog No. 5000349426
Encompass_Preferred
PP1 is a potent, Src family-selective tyrosine kinase inhibitor. It inhibits Lck and FynT in vitro at significantly lower concentrations than other kinases. It also inhibits whole cell tyrosine phosphorylation and proliferation in T cells, and selectively inhibits IL-2 gene expression.
  • Potent, Src family-selective tyrosine kinase inhibitor
  • Inhibits Lck with IC50 of 5 nM
  • Inhibits FynT with IC50 of 6 nM
  • Inhibits whole cell tyrosine phosphorylation
  • Selectively inhibits IL-2 gene expression
Catalog No. 50-003-49426 Supplier Medchemexpress LLC Supplier No. HY1380450MG
PP1 is a potent, Src family-selective tyrosine kinase inhibitor. It inhibits Lck and FynT in vitro at significantly lower concentrations than other kinases. It also inhibits whole cell tyrosine phosphorylation and proliferation in T cells, and selectively inhibits IL-2 gene expression.
  • Potent, Src family-selective tyrosine kinase inhibitor
  • Inhibits Lck with IC50 of 5 nM
  • Inhibits FynT with IC50 of 6 nM
  • Inhibits whole cell tyrosine phosphorylation
  • Selectively inhibits IL-2 gene expression