Learn More
STEMCELL Technologies PP1, Size: 1 mg
SDP
Supplier:  STEMCELL Technologies 73112
Encompass
Inhibit LCK, FYN, HCK, and SRC with PP1 (Hanke et al. J Biol Chem, 1996). This reversible inhibitor of the SRC family of tyrosine kinases regulates a variety of cellular processes, such as apoptosis and migration. It is used in cancer research to induce apoptosis in non-small cell lung cancer cell lines (Zhang et al. Am J Pathol, 2007) and block TGF-β-mediated migration of primary non-small cell lung carcinoma cells and pancreatic ductal adenocarcinoma cell lines (Bartscht et al. Cancer Chemother Pharmacol, 2012). PP1 is also used to reprogram mouse embryonic fibroblasts to induced pluripotent stem cells in the absence of reprogramming factor SOX2 (Staerk et al. Angew Chem Int Ed Engl, 2011; Ma et al. Circ Res, 2013). PP1 is supplied as a crystalline solid with ≥98% purity; Molecular weight 281.4 g/mol; CAS Number 172889-26-8. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-6028