Learn More
Cayman Chemical PP1 (Src Inhibitor), Purity: ≥98%, CAS: 172889-26-8, 10 mg
SDP
Catalog No. 502954430
Encompass_Preferred
A potent, reversible, ATP-competitive, and selective inhibitor of the Src family of protein tyrosine kinases: p56lck (IC50 = 5 nM), p59fynT (IC50 = 6 nM), Hck (IC50 = 20 nM)
Catalog No. 50-295-4430 Supplier Cayman Chemical Supplier No. 1424410 MG
A potent, reversible, ATP-competitive, and selective inhibitor of the Src family of protein tyrosine kinases: p56lck (IC50 = 5 nM), p59fynT (IC50 = 6 nM), Hck (IC50 = 20 nM)