Learn More
Medchemexpress LLC HY-13805 10mg , PP2 CAS:172889-27-9 Purity:>98%
SDP
Catalog No. 502260104
Encompass_Preferred
HY-13805 10mg PP2 CAS: 172889-27-9 PP2 is a reversible and ATP-competitive Src family kinases inhibitor with IC50s of 4 and 5 nM for Lck and Fyn, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0104 Supplier Medchemexpress LLC Supplier No. HY1380510MG
HY-13805 10mg PP2 CAS: 172889-27-9 PP2 is a reversible and ATP-competitive Src family kinases inhibitor with IC50s of 4 and 5 nM for Lck and Fyn, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.