Learn More
AdipoGen PP2
SDP
Catalog No. 502562287
Encompass_Preferred
Chemical. CAS 172889-27-9. Formula C15H16ClN5. MW 301.8. Highly potent and selective Scr family tyrosine kinase inhibitor. Reduces cancer metastasis. Apoptotic. Antitumor compound. RIP2 inhbitor. Blocks TGF-beta-mediated cellular responses. Autophagy modulator.
Catalog No. 50-256-2287 Supplier AdipoGen Supplier No. AGCR13563M001
Chemical. CAS 172889-27-9. Formula C15H16ClN5. MW 301.8. Highly potent and selective Scr family tyrosine kinase inhibitor. Reduces cancer metastasis. Apoptotic. Antitumor compound. RIP2 inhbitor. Blocks TGF-beta-mediated cellular responses. Autophagy modulator.