Learn More
Selleck Chemical LLC PP2 5mg 172889-27-9 AG 1879,AGL 1879
SDP
Catalog No. 501364318
Encompass_Preferred
PP2 (AG 1879, AGL 1879), a Src family kinase inhibitor, potently inhibits Lck/Fyn with IC50 of 4 nM/5 nM in cell-free assays, ~100-fold less potent to EGFR, inactive for ZAP-70, JAK2 and PKA. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4318 Supplier Selleck Chemical LLC Supplier No. S70085MG
Add to Cart
Add to Cart
PP2 (AG 1879, AGL 1879), a Src family kinase inhibitor, potently inhibits Lck/Fyn with IC50 of 4 nM/5 nM in cell-free assays, ~100-fold less potent to EGFR, inactive for ZAP-70, JAK2 and PKA. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.