Learn More
AdipoGen PP2 (5 mg)
SDP
Supplier:  AdipoGen AGCR13563M005
Encompass_Preferred
Chemical. CAS: 172889-27-9. Formula: C15H16ClN5. MW: 301.8. Highly potent and selective Scr family tyrosine kinase inhibitor. Reduces cancer metastasis. Apoptotic. Antitumor compound. RIP2 inhbitor. Blocks TGF-beta-mediated cellular responses. Autophagy modulator.
Catalog No. 50-205-3930