Learn More
Enzo Life Sciences Protein phosphatase 2 substrate (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP1520001
Encompass_Preferred
This peptide sequence, RRApTVA, has been found to be an excellent substrate for both PP2A and PP2C phosphatases. Alternative name: PP2 substrate. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-1260