Learn More
Enzo Life Sciences pp242 hydrate (1mg). CAS: 1092351-67-1
SDP
Catalog No. 502013424
Encompass_Preferred
Inhibitor of the active site of mTOR kinase in both mTORC1 and mTORC2. pp242 has been shown to cause the death of certain human and murine leukemia cells more potently than rapamycin. Alternative name: PP-242, 2-[4-Amino-1-(1-methylethyl)-1H-pyrazolo[3,4-d]pyrimidin-3-yl]-1H-indol-5-ol, 2-(4-amino-1-isopropyl-1H-pyrazolo[3,4-d]pyrimidin-3-yl)-1H-indol-5-ol. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.
Catalog No. 50-201-3424 Supplier Enzo Life Sciences Supplier No. ENZCHM1320001
Inhibitor of the active site of mTOR kinase in both mTORC1 and mTORC2. pp242 has been shown to cause the death of certain human and murine leukemia cells more potently than rapamycin. Alternative name: PP-242, 2-[4-Amino-1-(1-methylethyl)-1H-pyrazolo[3,4-d]pyrimidin-3-yl]-1H-indol-5-ol, 2-(4-amino-1-isopropyl-1H-pyrazolo[3,4-d]pyrimidin-3-yl)-1H-indol-5-ol. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.