Learn More
Selleck Chemical LLC Torkinib (PP242) 10mM/1mL 1092351-67-1
SDP
Supplier:  Selleck Chemical LLC S221810MM/1ML
Encompass_Preferred
Torkinib (PP242) is a selective mTOR inhibitor with IC50 of 8 nM in cell-free assays; targets both mTOR complexes with >10- and 100-fold selectivity for mTOR than PI3K? or PI3K?/?/?, respectively. Torkinib (PP242) induces mitophagy and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0551426
 Restricted Item