Learn More
Selleck Chemical LLC Torkinib (PP242) 50mg 1092351-67-1
SDP
Catalog No. 508771
Encompass_Preferred
Torkinib (PP242) is a selective mTOR inhibitor with IC50 of 8 nM in cell-free assays; targets both mTOR complexes with >10- and 100-fold selectivity for mTOR than PI3K? or PI3K?/?/?, respectively. Torkinib (PP242) induces mitophagy and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-877-1 Supplier Selleck Chemical LLC Supplier No. S221850MG
Torkinib (PP242) is a selective mTOR inhibitor with IC50 of 8 nM in cell-free assays; targets both mTOR complexes with >10- and 100-fold selectivity for mTOR than PI3K? or PI3K?/?/?, respectively. Torkinib (PP242) induces mitophagy and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.