Learn More
Aviva Systems Biology Corp PP2447 Antibody - N-terminal region (ARP42647_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42647_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PP2447. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PP2447. Alternate names: RP3-402G11.12, LP6054, MGC110928, PP2447ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-53347