Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PP2A alpha + beta Antibody (YA123) | Protein A affinity purified | 36 kDa | 100 UL
SDP

Catalog No. 5000397839
Encompass_Preferred

PP2A alpha + beta Antibody (YA123) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting to PP2A alpha + beta. It is for research use only.

  • Host: Rabbit; Clonality: Recombinant, monoclonal; Isotype: IgG
  • Molecular weight: 36 kDa
  • Species reactivity: Human, mouse, rat
  • Immunogen: Synthetic peptide for human PP2A alpha (AA 260-309)
  • Sensitivity: Endogenous
  • Purification: Protein A affinity purified
  • Conjugation: Non-conjugated
  • Appearance: Liquid
  • Formulation: 1*TBS (pH7.4), 0.05% BSA, 40% Glycerol, 0.05% Sodium Azide.
  • Storage: -20°C for 1 year. Avoid freeze/thaw cycles.
  • Shipping: Blue ice.
  • Applications: WB (1:1000-1:2000), ICC/IF (1:50-1:200), IHC-P (1:100-1:200), IP (assay dependent)

Catalog No. 50-003-97839 Supplier Medchemexpress LLC Supplier No. HYP80284100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PP2A alpha + beta Antibody (YA123) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting to PP2A alpha + beta. It is for research use only.

  • Host: Rabbit; Clonality: Recombinant, monoclonal; Isotype: IgG
  • Molecular weight: 36 kDa
  • Species reactivity: Human, mouse, rat
  • Immunogen: Synthetic peptide for human PP2A alpha (AA 260-309)
  • Sensitivity: Endogenous
  • Purification: Protein A affinity purified
  • Conjugation: Non-conjugated
  • Appearance: Liquid
  • Formulation: 1*TBS (pH7.4), 0.05% BSA, 40% Glycerol, 0.05% Sodium Azide.
  • Storage: -20°C for 1 year. Avoid freeze/thaw cycles.
  • Shipping: Blue ice.
  • Applications: WB (1:1000-1:2000), ICC/IF (1:50-1:200), IHC-P (1:100-1:200), IP (assay dependent)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.