Learn More
Enzo Life Sciences Protein Phosphatase 2A1 (bovine kidney), (purified) (1 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE1190001
Encompass_Preferred
Alternative name: PP2A1. Formulation: Liquid. In 50 mM Tris, pH 7.0, containing 0.1 mM EDTA, 50% glycerol, 14 mM 2-mercaptoethanol, 1 mM benzamidine, 0.1 mM PMSF. Source: Isolated from bovine kidney. Long Term Storage: -20°C.
Catalog No. 50-201-1936