Learn More
Sino Biological Recombinant Human PPP1CA Protein (GST Tag), Active
Catalog No. 502573868
Encompass_Preferred
Recombinant full length human PP1A was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-3868 Supplier Sino Biological Supplier No. P1320G500
Recombinant full length human PP1A was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.