Learn More
Abcam PP3, Negative control for PP2, 10MG
Catalog No. 502707419
Encompass_Preferred
MW 211.22 Da, Purity >97%. Negative control for the Src kinase inhibitor PP2 (ab120308). Inhibits EGFR kinase (IC₅₀ = 2.7 μM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7419 Supplier Abcam Supplier No. AB12061710MG
MW 211.22 Da, Purity >97%. Negative control for the Src kinase inhibitor PP2 (ab120308). Inhibits EGFR kinase (IC₅₀ = 2.7 μM).
The product is subject to the following: Abcam Restricted Use Statement